Gene Transcripts
- Gene Symbol
- 4930505M18Rik
- Entrez Gene ID
- 67641
- Gene Name
- RIKEN cDNA 4930505M18 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.72619
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015707.1 |
- |
1 |
95 |
|
AK019701.1 |
- |
1 |
68 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715465.1 |
- |
1 |
65 |
|
Chr. 7 - 37669827 - 37671549 [+] |
View Full Product Details