Gene Transcripts
- Gene Symbol
- LOC101929294
- Entrez Gene ID
- 101929294
- Gene Name
- uncharacterized LOC101929294
- Gene Aliases
- Chromosome Location
- Chr. 8: 24295814 - 24548618 on Build GRCh38
- UniGene ID
- Hs.637673
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_125808.1 |
|
6 |
836 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000518988.5 |
- |
4 |
690 |
|
Chr. 8 - 24295814 - 24387515 [-] |
View Full Product Details