Gene Transcripts
- Gene Symbol
- LY96
- Entrez Gene ID
- 23643
- Gene Name
- lymphocyte antigen 96
- Gene Aliases
- ESOP-1, MD-2, MD2, ly-96
- Chromosome Location
- Chr. 8: 73991275 - 74099807 on Build GRCh38
- UniGene ID
- Hs.726603
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000519323.1 |
- |
2 |
588 |
|
Chr. 8 - 74052340 - 74099853 [+] |
GenBankmRNA |
BC022402.1 |
- |
2 |
588 |
|
Chr. 8 - 74052340 - 74099853 [+] |
View Full Product Details