Gene Transcripts

Gene Symbol
2310069B03Rik
Entrez Gene ID
69652
Gene Name
RIKEN cDNA 2310069B03 gene
Gene Aliases
Chromosome Location
Chr. 6: 82877866 - 82881853 on Build GRCm38
UniGene ID
Mm.96357
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040520.1 4 866
GenBank mRNA AK010122.1 - 4 881
AK078808.1 - 4 867

View Full Product Details