Gene Transcripts

Gene Symbol
Zmynd10
Entrez Gene ID
114602
Gene Name
zinc finger, MYND domain containing 10
Gene Aliases
Blu
Chromosome Location
Chr. 9: 107545991 - 107551321 on Build GRCm38
UniGene ID
Mm.28331
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053253.3 NP_444483.2 6 722
GenBank mRNA AK132332.1 - 6 731
BC052357.1 - 6 722 6772144

View Full Product Details