Gene Transcripts

Gene Symbol
Ginm1
Entrez Gene ID
215751
Gene Name
glycoprotein integral membrane 1
Gene Aliases
AA589616
Chromosome Location
Chr. 10: 7767947 - 7780946 on Build GRCm38
UniGene ID
Mm.471825
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145418.4 NP_663393.1 3 243
GenBank mRNA AK144481.1 - 3 204
BC013529.1 - 3 243 4238526

View Full Product Details