Gene Transcripts

Gene Symbol
Olfr1336
Entrez Gene ID
258917
Gene Name
olfactory receptor 1336
Gene Aliases
MOR103-7
Chromosome Location
Chr. 7: 6460511 - 6461455 on Build GRCm38
UniGene ID
Mm.223322
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146915.1 NP_667126.1 1 335
GenBank mRNA BC104285.1 - 1 334
BC104286.1 - 1 335 40042538

View Full Product Details