Gene Transcripts

Gene Symbol
Olfr1257
Entrez Gene ID
258984
Gene Name
olfactory receptor 1257
Gene Aliases
MOR232-1
Chromosome Location
Chr. 2: 89880828 - 89881757 on Build GRCm38
UniGene ID
Mm.389309
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146982.1 NP_667193.1 1 810
GenBank mRNA BC137779.1 - 1 844

View Full Product Details