Gene Transcripts

Gene Symbol
Hnrnph3
Entrez Gene ID
432467
Gene Name
heterogeneous nuclear ribonucleoprotein H3
Gene Aliases
AA693301, AI666703, Hnrph3
Chromosome Location
Chr. 10: 63014650 - 63023889 on Build GRCm38
UniGene ID
Mm.274784
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079824.1 NP_001073293.1 8, 9 1021
GenBank mRNA BC147496.1 - 6, 7 678
BX514270.1 - 3, 4 250

View Full Product Details