Gene Transcripts
- Gene Symbol
- Lta
- Entrez Gene ID
- 25008
- Gene Name
- lymphotoxin alpha
- Gene Aliases
- Tnfb
- Chromosome Location
- Chr. 20: 4852671 - 4854677 on Build Rnor_6.0
- UniGene ID
- Rn.160577
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080769.2 |
NP_542947.1
|
4 |
635 |
|
- Gene Symbol
- LOC103690381
- Entrez Gene ID
- 103690381
- Gene Name
- lymphotoxin-alpha-like
- Gene Aliases
- Chromosome Location
- Chr. 20: 5193153 - 5194125 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details