Gene Transcripts

Gene Symbol
Irs1
Entrez Gene ID
25467
Gene Name
insulin receptor substrate 1
Gene Aliases
IRS1IRM
Chromosome Location
Chr. 9: 88033668 - 88086488 on Build Rnor_6.0
UniGene ID
Rn.10476
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012969.1 NP_037101.1 1 3814
GenBank mRNA X58375.1 - 1 3814

View Full Product Details