Gene Transcripts

Gene Symbol
Fgf17
Entrez Gene ID
29368
Gene Name
fibroblast growth factor 17
Gene Aliases
Chromosome Location
Chr. 15: 52326769 - 52337332 on Build Rnor_6.0
UniGene ID
Rn.44353
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019198.1 NP_062071.1 5 389
XM_008770827.1 XP_008769049.1 6 545
GenBank mRNA AB008682.1 - 5 389

View Full Product Details