Gene Transcripts

Gene Symbol
Zfp148
Entrez Gene ID
58820
Gene Name
zinc finger protein 148
Gene Aliases
Zbp-89, Znf148
Chromosome Location
Chr. 11: 70510573 - 70618337 on Build Rnor_6.0
UniGene ID
Rn.64671
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031615.1 NP_113803.1 4 574
XM_006248435.2 XP_006248497.1 1 204
GenBank mRNA U30381.1 - 4 574

View Full Product Details