Gene Transcripts

Gene Symbol
Fat1
Entrez Gene ID
83720
Gene Name
FAT atypical cadherin 1
Gene Aliases
Fat, Fath
Chromosome Location
Chr. 16: 50372150 - 50501716 on Build Rnor_6.0
UniGene ID
Rn.8633
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031819.2 NP_114007.1 10 5323
XM_008771269.1 XP_008769491.1 8 1950
GenBank mRNA AF100960.1 - 10 5325

View Full Product Details