Gene Transcripts

Gene Symbol
SYN1
Entrez Gene ID
6853
Gene Name
synapsin I
Gene Aliases
SYN1a, SYN1b, SYNI
Chromosome Location
Chr. X: 47571901 - 47619857 on Build GRCh38
UniGene ID
Hs.225936
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006950.3 NP_008881.2 5, 6 910
NM_133499.2 NP_598006.1 5, 6 910

View Full Product Details