Gene Transcripts

Gene Symbol
F11r
Entrez Gene ID
116479
Gene Name
F11 receptor
Gene Aliases
Jam1
Chromosome Location
Chr. 13: 89826272 - 89850151 on Build Rnor_6.0
UniGene ID
Rn.107273
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053796.1 NP_446248.1 7 860
GenBank mRNA AF241261.1 - 3 407
AF276998.1 - 7 860
BC065309.1 - 7 842 5599898

View Full Product Details