Gene Transcripts

Gene Symbol
Pdk1
Entrez Gene ID
116551
Gene Name
pyruvate dehydrogenase kinase, isozyme 1
Gene Aliases
Chromosome Location
Chr. 3: 58530870 - 58561493 on Build Rnor_6.0
UniGene ID
Rn.11185
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053826.2 NP_446278.2 2, 3 443
XM_006234352.2 XP_006234414.1 2, 3 215
GenBank mRNA BC089783.1 - 2, 3 443
L22294.1 - 2, 3 374

View Full Product Details