Gene Transcripts

Gene Symbol
Slc12a9
Entrez Gene ID
171443
Gene Name
solute carrier family 12, member 9
Gene Aliases
Ccc6, Cip1
Chromosome Location
Chr. 12: 22434845 - 22451265 on Build Rnor_6.0
UniGene ID
Rn.176567
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134405.1 NP_599232.1 2 266
GenBank mRNA AB023645.1 - 2 266

View Full Product Details