Gene Transcripts

Gene Symbol
Man2c1
Entrez Gene ID
246136
Gene Name
mannosidase, alpha, class 2C, member 1
Gene Aliases
Chromosome Location
Chr. 8: 61805697 - 61816800 on Build Rnor_6.0
UniGene ID
Rn.11301
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139256.1 NP_640349.1 8 1058
GenBank mRNA BC086989.1 - 8 939
M57547.1 - 8 1058

View Full Product Details