Gene Transcripts
- Gene Symbol
- Olr1413
- Entrez Gene ID
- 252887
- Gene Name
- olfactory receptor 1413
- Gene Aliases
- Olfr30, TPCR05
- Chromosome Location
- Chr. 10: 43894429 - 43895376 on Build Rnor_6.0
- UniGene ID
- Rn.92442
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_214821.1 |
NP_999986.1
|
1 |
776 |
|
GenBank mRNA |
X89695.1 |
- |
1 |
399 |
|
View Full Product Details