Gene Transcripts

Gene Symbol
Soat2
Entrez Gene ID
266770
Gene Name
sterol O-acyltransferase 2
Gene Aliases
Acat-2
Chromosome Location
Chr. 7: 143754892 - 143767989 on Build Rnor_6.0
UniGene ID
Rn.83628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153728.2 NP_714950.2 13 1634
GenBank mRNA AB075946.1 - 13 1366
AB101480.1 - 13 1634

View Full Product Details