Gene Transcripts
- Gene Symbol
- TMSB4X
- Entrez Gene ID
- 7114
- Gene Name
- thymosin beta 4, X-linked
- Gene Aliases
- FX, PTMB4, TB4X, TMSB4
- Chromosome Location
- Chr. X: 12975107 - 12977227 on Build GRCh38
- UniGene ID
- Hs.437277
- Species
- Homo sapiens
- Gene Symbol
- TMSB4XP1
- Entrez Gene ID
- 7115
- Gene Name
- thymosin beta 4, X-linked pseudogene 1
- Gene Aliases
- TMSL1
- Chromosome Location
- Chr. 1: 42499781 - 42500417 on Build GRCh38
- UniGene ID
- Hs.655548
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC090885.1 |
- |
1 |
181 |
|
View Full Product Details