Gene Transcripts

Gene Symbol
Poglut1
Entrez Gene ID
288091
Gene Name
protein O-glucosyltransferase 1
Gene Aliases
Ktelc1, RGD1306248
Chromosome Location
Chr. 11: 64761493 - 64788210 on Build Rnor_6.0
UniGene ID
Rn.7051
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100652.1 NP_001094122.1 3, 4 425
GenBank mRNA BC091408.1 - 2, 3 197

View Full Product Details