Gene Transcripts

Gene Symbol
Hoxd3
Entrez Gene ID
288152
Gene Name
homeo box D3
Gene Aliases
Hox4r6
Chromosome Location
Chr. 3: 61650588 - 61670285 on Build Rnor_6.0
UniGene ID
Rn.223085
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271038.1 NP_001257967.1 3 1296
XM_008761880.1 XP_008760102.1 2 1556
XM_008761881.1 XP_008760103.1 3 1034

View Full Product Details