Gene Transcripts

Gene Symbol
Olr1605
Entrez Gene ID
289923
Gene Name
olfactory receptor 1605
Gene Aliases
ratchr15-12385736-12384768_ORF
Chromosome Location
Chr. 15: 12128252 - 12129220 on Build Rnor_6.0
UniGene ID
Rn.142463
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000088.1 NP_001000088.1 1 291

View Full Product Details