Gene Transcripts

Gene Symbol
Pak4
Entrez Gene ID
292756
Gene Name
p21 protein (Cdc42/Rac)-activated kinase 4
Gene Aliases
Chromosome Location
Chr. 1: 85162462 - 85201746 on Build Rnor_6.0
UniGene ID
Rn.18543
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106238.1 NP_001099708.1 6 1405
XM_008759119.1 XP_008757341.1 6 3455
GenBank mRNA BC168940.1 - 6 1415

View Full Product Details