Gene Transcripts

Gene Symbol
Cpeb1
Entrez Gene ID
293056
Gene Name
cytoplasmic polyadenylation element binding protein 1
Gene Aliases
Chromosome Location
Chr. 1: 143171457 - 143278485 on Build Rnor_6.0
UniGene ID
Rn.224485
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106276.1 NP_001099746.1 10 1474
XM_008759509.1 XP_008757731.1 10 2083
XM_008759510.1 XP_008757732.1 10 2068

View Full Product Details