Gene Transcripts

Gene Symbol
Dnajc10
Entrez Gene ID
295690
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 10
Gene Aliases
Chromosome Location
Chr. 3: 67538289 - 67578308 on Build Rnor_6.0
UniGene ID
Rn.8642
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106486.2 NP_001099956.2 13, 14 1576
GenBank mRNA BC100105.1 - 6, 7 532
FQ226920.1 - 12, 13 1187

View Full Product Details