Gene Transcripts

Gene Symbol
Olr1251
Entrez Gene ID
300576
Gene Name
olfactory receptor 1251
Gene Aliases
ratchr8-40765174-40766103_ORF
Chromosome Location
Chr. 4: 1671269 - 1672198 on Build Rnor_6.0
UniGene ID
Rn.143519
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000453.1 NP_001000453.1 1 89

View Full Product Details