Gene Transcripts

Gene Symbol
Nkx2-3
Entrez Gene ID
309389
Gene Name
NK2 homeobox 3
Gene Aliases
Chromosome Location
Chr. 1: 263361426 - 263371043 on Build Rnor_6.0
UniGene ID
Rn.91986
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107594.1 NP_001101064.1 1 205
XM_008760427.1 XP_008758649.1 2 1425

View Full Product Details