Gene Transcripts

Gene Symbol
Mcm2
Entrez Gene ID
312538
Gene Name
minichromosome maintenance complex component 2
Gene Aliases
Chromosome Location
Chr. 4: 120825699 - 120840221 on Build Rnor_6.0
UniGene ID
Rn.2715
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107873.2 NP_001101343.2 14, 15 2489
XM_006236863.1 XP_006236925.1 14, 15 2599
GenBank mRNA FQ220778.1 - 2, 3 282

View Full Product Details