Gene Transcripts

Gene Symbol
MGC114427
Entrez Gene ID
317297
Gene Name
similar to melanoma antigen family A, 5
Gene Aliases
Chromosome Location
Chr. X: 147491321 - 147496650 on Build Rnor_6.0
UniGene ID
Rn.54113
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024893.1 NP_001020064.1 3 467
XM_008773565.1 XP_008771787.1 4 803
GenBank mRNA BC097387.1 - 3 467

View Full Product Details