Gene Transcripts

Gene Symbol
Nkd1
Entrez Gene ID
364952
Gene Name
naked cuticle homolog 1 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 19: 19434731 - 19509087 on Build Rnor_6.0
UniGene ID
Rn.8826
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271381.1 NP_001258310.1 3, 4 161
XM_008772413.1 XP_008770635.1 2, 3 326
GenBank mRNA BC168970.1 - 2, 3 327

View Full Product Details