Gene Transcripts

Gene Symbol
Slc7a14
Entrez Gene ID
499587
Gene Name
solute carrier family 7, member 14
Gene Aliases
Chromosome Location
Chr. 2: 115678344 - 115788688 on Build Rnor_6.0
UniGene ID
Rn.44848
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134615.1 NP_001128087.1 8 2491

View Full Product Details