Gene Transcripts

Gene Symbol
Ndufb10
Entrez Gene ID
681418
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10
Gene Aliases
Chromosome Location
Chr. 10: 14090128 - 14092289 on Build Rnor_6.0
UniGene ID
Rn.2867
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109443.1 NP_001102913.1 2, 3 380
GenBank mRNA FQ218220.1 - 2, 3 325

View Full Product Details