Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008838.1 | NP_032864.1 | 3 | 417 | |
GenBank mRNA | AK132796.1 | - | 3 | 423 | |
AK171083.1 | - | 3 | 405 | ||
AK218842.1 | - | 3 | 329 | ||
BC028862.1 | - | 3 | 406 | 3673399 | |
D50264.1 | - | 3 | 417 |