Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001104546.1 | NP_001098016.1 | 6 | 420 | |
NM_006413.4 | NP_006404.1 | 6 | 420 | ||
XM_011539120.2 | XP_011537422.1 | 9 | 973 | ||
XM_011539121.2 | XP_011537423.1 | 9 | 973 | ||
XM_017015475.1 | XP_016870964.1 | 9 | 973 | ||
XM_017015476.1 | XP_016870965.1 | 9 | 973 | ||
XM_017015477.1 | XP_016870966.1 | 7 | 673 | ||
XM_017015478.1 | XP_016870967.1 | 9 | 973 | ||
GenBank mRNA | AK225532.1 | - | 6 | 393 | |
AK296196.1 | - | 6 | 398 | ||
AK312900.1 | - | 6 | 394 | ||
AL833628.1 | - | 6 | 406 | ||
BC006991.1 | - | 7 | 679 | 3827681 | |
DB049454.1 | - | 6 | 420 | ||
U77665.1 | - | 6 | 412 |