Gene Transcripts

Gene Symbol
Krt9
Entrez Gene ID
266717
Gene Name
keratin 9, type I
Gene Aliases
Krt1-9
Chromosome Location
Chr. 10: 88102519 - 88107232 on Build Rnor_6.0
UniGene ID
Rn.145135
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153476.1 NP_703206.1 3 788
GenBank mRNA AY128946.1 - 3 788

View Full Product Details