Gene Transcripts

Gene Symbol
Vwa7
Entrez Gene ID
309611
Gene Name
von Willebrand factor A domain containing 7
Gene Aliases
C6orf27, G7c, NG37
Chromosome Location
Chr. 20: 5008362 - 5019040 on Build Rnor_6.0
UniGene ID
Rn.219144
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212499.2 NP_997664.1 14 2127
XM_008772728.1 XP_008770950.1 13 2561

View Full Product Details