Gene Transcripts
- Gene Symbol
- Trpm1
- Entrez Gene ID
- 17364
- Gene Name
- transient receptor potential cation channel, subfamily M, member 1
- Gene Aliases
- 4732499L03Rik, AI606771, Ltrpc1, Mlsn1
- Chromosome Location
- Chr. 7: 64153835 - 64269772 on Build GRCm38
- UniGene ID
- Mm.297430
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039104.2 |
NP_001034193.2
|
7, 8 |
1231 |
|
NM_018752.3 |
NP_061222.3
|
7, 8 |
1124 |
|
GenBank mRNA |
AF047714.1 |
- |
7, 8 |
993 |
|
AK044507.1 |
- |
7, 8 |
1222 |
|
AK148094.1 |
- |
7, 8 |
1029 |
|
AK148193.1 |
- |
7, 8 |
1032 |
|
AK148268.1 |
- |
7, 8 |
978 |
|
AK148275.1 |
- |
7, 8 |
1034 |
|
AK149290.1 |
- |
7, 8 |
1153 |
|
AK164395.1 |
- |
7, 8 |
1034 |
|
AK164423.1 |
- |
7, 8 |
1041 |
|
AK191785.1 |
- |
1, 2 |
120 |
|
AK192880.1 |
- |
|
222 |
|
AK197227.1 |
- |
|
134 |
|
AK203068.1 |
- |
1, 2 |
47 |
|
AK208031.1 |
- |
1, 2 |
120 |
|
AK213026.1 |
- |
1, 2 |
47 |
|
AK214278.1 |
- |
1, 2 |
120 |
|
AK216151.1 |
- |
1, 2 |
129 |
|
AK217386.1 |
- |
1, 2 |
121 |
|
AY180104.2 |
- |
7, 8 |
1231 |
|
BC082560.1 |
- |
7, 8 |
1000 |
|
BC085168.1 |
- |
6, 7 |
803 |
|
View Full Product Details