Gene Transcripts

Gene Symbol
Krtap5-1
Entrez Gene ID
50774
Gene Name
keratin associated protein 5-1
Gene Aliases
Chromosome Location
Chr. 7: 142296377 - 142297069 on Build GRCm38
UniGene ID
Mm.389993
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015808.1 NP_056623.1 1 531
GenBank mRNA BC104355.1 - 417

View Full Product Details