Gene Transcripts

Gene Symbol
Krtap5-1
Entrez Gene ID
50774
Gene Name
keratin associated protein 5-1
Gene Aliases
Chromosome Location
Chr. 7: 142296377 - 142297069 on Build GRCm38
UniGene ID
Mm.389993
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015808.1 NP_056623.1 1 158
GenBank mRNA BC104355.1 - 158

Gene Symbol
Gm4559
Entrez Gene ID
100043627
Gene Name
predicted gene 4559
Gene Aliases
Chromosome Location
Chr. 7: 142273764 - 142274363 on Build GRCm38
UniGene ID
Mm.388589
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199309.1 NP_001186238.1 1 95
GenBank mRNA BC104354.1 - 1 95
BC120878.1 - 1 94

View Full Product Details