Gene Transcripts

Gene Symbol
Slc41a1
Entrez Gene ID
363985
Gene Name
solute carrier family 41 (magnesium transporter), member 1
Gene Aliases
Chromosome Location
Chr. 13: 48607308 - 48627999 on Build Rnor_6.0
UniGene ID
Rn.145261
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108855.1 NP_001102325.1 11 2415
XM_008769486.1 XP_008767708.1 10 1538

View Full Product Details