Gene Transcripts

Gene Symbol
Cpne3
Entrez Gene ID
313087
Gene Name
copine III
Gene Aliases
Chromosome Location
Chr. 5: 33525307 - 33574608 on Build Rnor_6.0
UniGene ID
Rn.4034
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107917.2 NP_001101387.2 2 157
XM_006237932.1 XP_006237994.1 3 279
GenBank mRNA AM746508.1 - 2 157

View Full Product Details