Gene Transcripts

Gene Symbol
CNGA2
Entrez Gene ID
1260
Gene Name
cyclic nucleotide gated channel alpha 2
Gene Aliases
CNCA, CNCA1, CNG2, OCNC1, OCNCALPHA, OCNCa
Chromosome Location
Chr. X: 151734746 - 151745564 on Build GRCh38
UniGene ID
Hs.447360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005140.1 NP_005131.1 7 1032
GenBank mRNA AK128186.1 - 7 1032
BC126302.1 - 7 901 8992018
BC126304.1 - 7 901 8992020

View Full Product Details