Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025190.3 | NP_079466.3 | 39 | 3501 | |
XM_005263995.4 | XP_005264052.1 | 39 | 3781 | ||
XM_006712661.1 | XP_006712724.1 | 39 | 3781 | ||
XM_017004578.1 | XP_016860067.1 | 39 | 3778 | ||
XM_017004579.1 | XP_016860068.1 | 37 | 3679 | ||
XM_017004580.1 | XP_016860069.1 | 37 | 3679 | ||
XM_017004581.1 | XP_016860070.1 | 37 | 3679 | ||
XM_017004582.1 | XP_016860071.1 | 37 | 3679 | ||
XM_017004583.1 | XP_016860072.1 | 38 | 3643 | ||
XM_017004584.1 | XP_016860073.1 | 33 | 3475 | ||
XM_017004585.1 | XP_016860074.1 | 37 | 3536 | ||
XM_017004586.1 | XP_016860075.1 | 34 | 2522 | ||
XM_017004587.1 | XP_016860076.1 | 38 | 3710 | ||
XM_017004588.1 | XP_016860077.1 | 37 | 3575 | ||
XM_017004589.1 | XP_016860078.1 | 36 | 3468 | ||
XM_017004590.1 | XP_016860079.1 | 35 | 3395 | ||
XM_017004591.1 | XP_016860080.1 | 21 | 3499 | ||
XM_017004592.1 | XP_016860081.1 | 40 | 3825 | ||
XM_017004593.1 | XP_016860082.1 | 38 | 3631 | ||
XM_017004594.1 | XP_016860083.1 | 37 | 3524 | ||
XM_017004595.1 | XP_016860084.1 | 38 | 3580 |