Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022894.3 | NP_075045.2 | 19 | 2206 | |
XM_005264500.3 | XP_005264557.1 | 19 | 2206 | ||
XM_005264501.1 | XP_005264558.1 | 19 | 2024 | ||
XM_011533041.1 | XP_011531343.1 | 18 | 1912 | ||
XM_017004723.1 | XP_016860212.1 | 18 | 2068 | ||
XM_017004724.1 | XP_016860213.1 | 18 | 2068 | ||
GenBank mRNA | AB209304.1 | - | 9 | 961 | |
AF312211.1 | - | 19 | 2188 | ||
AJ308101.1 | - | 19 | 1957 | ||
AK023034.1 | - | 8 | 908 | ||
AK023544.1 | - | 18 | 1955 | ||
AK024249.1 | - | 7 | 823 | ||
AK314663.1 | - | 19 | 2170 | ||
AY029162.1 | - | 19 | 1966 | ||
BC111701.1 | - | 19 | 2080 | 40038554 | |
BC113747.1 | - | 19 | 2080 | 40038563 | |
BX647824.1 | - | 18 | 1926 |