Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282150.1 | NP_001269079.1 | 6 | 722 | |
NM_001282151.1 | NP_001269080.1 | 3 | 331 | ||
NM_016226.4 | NP_057310.1 | 4 | 567 | ||
NM_057180.2 | NP_476528.1 | 5 | 579 | ||
NR_104099.1 | 6 | 619 | |||
NR_104100.1 | 5 | 607 | |||
NR_104101.1 | 5 | 657 | |||
NR_104102.1 | 4 | 458 | |||
XM_006719459.3 | XP_006719522.1 | 6 | 628 | ||
XM_006719460.3 | XP_006719523.1 | 7 | 665 | ||
XM_011538484.2 | XP_011536786.1 | 5 | 613 | ||
XM_017019457.1 | XP_016874946.1 | 5 | 638 | ||
XM_017019458.1 | XP_016874947.1 | 4 | 758 | ||
XM_017019459.1 | XP_016874948.1 | 4 | 420 | ||
GenBank mRNA | AA148059.1 | - | 4 | 404 | |
AB578890.1 | - | 3 | 306 | ||
AF168716.1 | - | 5 | 537 | ||
AF175264.1 | - | 4 | 536 | ||
AF193795.1 | - | 4 | 529 | ||
AF201936.1 | - | 5 | 624 | ||
AF201946.1 | - | 4 | 515 | ||
AK000499.1 | - | 3 | 295 | ||
AK129767.1 | - | 4 | 529 | ||
AK130510.1 | - | 1365 | |||
AL136614.1 | - | 4 | 545 | ||
AL832866.1 | - | 4 | 2560 | ||
BC000880.1 | - | 5 | 537 | 3461977 | |
BC015095.1 | - | 5 | 546 | ||
BC095446.1 | - | 4 | 538 | 30719438 | |
BF030144.1 | - | 579 | |||
BG707325.1 | - | 4 | 562 | ||
BM806646.1 | - | 6 | 675 | ||
BU568689.1 | - | 6 | 580 | ||
CB158821.1 | - | 3 | 265 | ||
CR457182.1 | - | 3 | 490 | ||
CR533468.1 | - | 3 | 478 |