Gene Transcripts
- Gene Symbol
- TMTC1
- Entrez Gene ID
- 83857
- Gene Name
- transmembrane and tetratricopeptide repeat containing 1
- Gene Aliases
- ARG99, OLF, TMTC1A
- Chromosome Location
- Chr. 12: 29500813 - 29784759 on Build GRCh38
- UniGene ID
- Hs.401954
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001193451.1 |
NP_001180380.1
|
8, 9 |
1485 |
|
NM_175861.3 |
NP_787057.2
|
8, 9 |
1576 |
|
XM_005253499.4 |
XP_005253556.1
|
10, 11 |
1737 |
|
XM_017020004.1 |
XP_016875493.1
|
9, 10 |
1530 |
|
XM_017020005.1 |
XP_016875494.1
|
7, 8 |
1343 |
|
XM_017020006.1 |
XP_016875495.1
|
10, 11 |
1741 |
|
XM_017020007.1 |
XP_016875496.1
|
6, 7 |
763 |
|
GenBank mRNA |
AK055962.1 |
- |
6, 7 |
737 |
|
AK123619.1 |
- |
4, 5 |
1167 |
|
AK127297.1 |
- |
10, 11 |
1547 |
|
BC042083.1 |
- |
8, 9 |
1576 |
5749757 |
GQ865601.1 |
- |
8, 9 |
1485 |
|
View Full Product Details