Gene Transcripts

Gene Symbol
CEP41
Entrez Gene ID
95681
Gene Name
centrosomal protein 41
Gene Aliases
JBTS15, TSGA14
Chromosome Location
Chr. 7: 130393771 - 130441210 on Build GRCh38
UniGene ID
Hs.368315
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257158.1 NP_001244087.1 8 844
NM_001257159.1 NP_001244088.1 7 796
NM_018718.2 NP_061188.1 8 844
NR_046443.1 8 768
XM_011516709.2 XP_011515011.1 9 805
XM_011516710.2 XP_011515012.1 7 620
GenBank mRNA AF429308.1 - 8 690
AJ278890.1 - 8 647
AK026098.1 - 8 644
AK298618.1 - 7 612
AK311382.1 - 8 585
AK314676.1 - 8 623
AL359617.1 - 2 145
BC018789.1 - 8 626
BC056162.1 - 8 617 6142659

View Full Product Details